Learn More
Fireflysci Type 96 X-Rite Colorimeter Cell (Material: Optical Glass) (Lightpath: 20mm)
SDP
Catalog No. 501060326
Encompass_Preferred
Type 96 X-Rite Colorimeter Cell (Material: Optical Glass) (Lightpath: 20mm)• Cuvette has an open top• Is a standard cell for use with X-Rite colorimeters • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 20mm• Outside dimensions 26x28x40mm• Inside Dimensions- 20x24x40mm• Capacity- 14mL
Catalog No. 50-106-0326 Supplier Fireflysci Supplier No. 96G20
Type 96 X-Rite Colorimeter Cell (Material: Optical Glass) (Lightpath: 20mm)• Cuvette has an open top• Is a standard cell for use with X-Rite colorimeters • Material is Optical Glass (transmits over 80% between 340-2,500nm)• Two windows optically polished• Lightpath is 20mm• Outside dimensions 26x28x40mm• Inside Dimensions- 20x24x40mm• Capacity- 14mL