Learn More
Fireflysci Type 9FLP Disposable Semi-Micro Fluorescence Cuvettes (Lightpath: 10mm) (Material: PMMA)
SDP
Catalog No. 501060450
Encompass_Preferred
Type 9FLP Disposable Semi-Micro Fluorescence Cuvettes (Lightpath: 10mm) (Material: PMMA)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is PMMA (transmits over 80% between 285-900nm)• Four windows and the base optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x4x45mm• Capacity- 1.5mL
Catalog No. 50-106-0450 Supplier Fireflysci Supplier No. 9FLPMMA
Type 9FLP Disposable Semi-Micro Fluorescence Cuvettes (Lightpath: 10mm) (Material: PMMA)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is PMMA (transmits over 80% between 285-900nm)• Four windows and the base optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x4x45mm• Capacity- 1.5mL