Learn More
Fireflysci Type 9P Disposable Semi-Micro Cuvettes (Lightpath: 10mm) (Material: UV Plastic)
SDP
Catalog No. 501060449
Encompass_Preferred
Type 9P Disposable Semi-Micro Cuvettes (Lightpath: 10mm) (Material: UV Plastic)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is UV Plastic (transmits over 80% between 200-900nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x4x45mm• Capacity- 1.5mL
Catalog No. 50-106-0449 Supplier Fireflysci Supplier No. 9PUV
Type 9P Disposable Semi-Micro Cuvettes (Lightpath: 10mm) (Material: UV Plastic)• Cuvette has an open top with optional push caps that fit into the top (part # for the caps is PLCAPS100)• Comes in a box of 100• Material is UV Plastic (transmits over 80% between 200-900nm)• Two windows optically polished• Lightpath is 10mm• Outside dimensions 12.5x12.5x45mm• Inside Dimensions- 10x4x45mm• Capacity- 1.5mL