Learn More
Fireflysci A20 Cuvette Mount with Extended Spring (For Lightpaths Below 5mm)
SDP
Catalog No. 501060493
Encompass
Cuvette mount for rectangular cells with a lightpath of less than 5mm• This mount enables short pathlength cuvettes to fit in a 10mm cuvette holder• Contains an extended spring which makes it easier to remove from a spectrophotometer cell holder• Compatible with several models of FireflySci cuvettes- Type 1 (1 and 2mm lightpath version), Type 19, Type 20, Type 21 (0.5mm, 1mm, and 2mm versions), Type 48, Type 49
Catalog No. 50-106-0493 Supplier Fireflysci Supplier No. A20ES
Cuvette mount for rectangular cells with a lightpath of less than 5mm• This mount enables short pathlength cuvettes to fit in a 10mm cuvette holder• Contains an extended spring which makes it easier to remove from a spectrophotometer cell holder• Compatible with several models of FireflySci cuvettes- Type 1 (1 and 2mm lightpath version), Type 19, Type 20, Type 21 (0.5mm, 1mm, and 2mm versions), Type 48, Type 49