Learn More
Fireflysci A25 Cuvette Spacer (For Lightpaths of 2mm)
SDP
Catalog No. 501060497
Encompass
Cuvette spacer for rectangular cuvettes a lightpath of 1mm to enable it to fit in a standard 10mm cuvette holder• A25 cuvette spacers are made from high quality black anodized aluminum• List of compatible FireflySci cuvettes: Type 1 (1mm version), Type 1FL (1mm version), Type 11 (1mm version), Type 21 (1mm version), Type 30 (1mm version), Type 30BM (1mm version), Type 41 (1mm version), Type 61 (1mm version)
Catalog No. 50-106-0497 Supplier Fireflysci Supplier No. A25LP2
Cuvette spacer for rectangular cuvettes a lightpath of 1mm to enable it to fit in a standard 10mm cuvette holder• A25 cuvette spacers are made from high quality black anodized aluminum• List of compatible FireflySci cuvettes: Type 1 (1mm version), Type 1FL (1mm version), Type 11 (1mm version), Type 21 (1mm version), Type 30 (1mm version), Type 30BM (1mm version), Type 41 (1mm version), Type 61 (1mm version)