Learn More
Fireflysci A25 Cuvette Spacer (For Lightpaths of 5mm)
SDP
Catalog No. 501060498
Encompass
Cuvette spacer for rectangular cuvettes a lightpath of 5mm to enable it to fit in a standard 10mm cuvette holder• A25 cuvette spacers are made from high quality black anodized aluminum• List of compatible FireflySci cuvettes: Type 1 (5mm version), Type 1FL (5mm version), Type 9 (5mm version), Type 9M (5mm version), Type 11 (5mm version), Type 11FL (5mm version), Type 21 (5mm version), Type 21FL (5mm version), Type 29 (5mm version), Type 29BM (5mm version), Type 29M (5mm version) Type 30 (5mm version), Type 30BM (5mm version), Type 30M (5mm version), Type 41 (5mm version), Type 41FL (5mm version), Type 61 (5mm version)
Catalog No. 50-106-0498 Supplier Fireflysci Supplier No. A25LP5
Cuvette spacer for rectangular cuvettes a lightpath of 5mm to enable it to fit in a standard 10mm cuvette holder• A25 cuvette spacers are made from high quality black anodized aluminum• List of compatible FireflySci cuvettes: Type 1 (5mm version), Type 1FL (5mm version), Type 9 (5mm version), Type 9M (5mm version), Type 11 (5mm version), Type 11FL (5mm version), Type 21 (5mm version), Type 21FL (5mm version), Type 29 (5mm version), Type 29BM (5mm version), Type 29M (5mm version) Type 30 (5mm version), Type 30BM (5mm version), Type 30M (5mm version), Type 41 (5mm version), Type 41FL (5mm version), Type 61 (5mm version)