Learn More
Fireflysci Type P66 Cell Holder- Holds 12 10mm Cuvettes
SDP
Catalog No. 501060491
Encompass_Preferred
Cell holder rack for holding 12 standard size 10mm pathlength cuvettes• Made of polyethylene so that the windows don't get scratched
Catalog No. 50-106-0491 Supplier Fireflysci Supplier No. P66
Add to Cart
Add to Cart
Cell holder rack for holding 12 standard size 10mm pathlength cuvettes• Made of polyethylene so that the windows don't get scratched