Learn More
Fireflysci P69 Mixer for 10mm Macro Cuvettes (Type 1, Type 3)
SDP
Catalog No. 501060510
Encompass_Preferred
Manual mixer fits any standard 10mm pathlength cuveette with a 10mm inside width• Made of clear acrylic with a straight handle, this tool is used when reactives are performed directly into cuvettes to mix added reagents. Up and down motion in cell provides complete mixing• Shaft diameter- 3.17mm (0.125 inches)
Catalog No. 50-106-0510 Supplier Fireflysci Supplier No. P69
Manual mixer fits any standard 10mm pathlength cuveette with a 10mm inside width• Made of clear acrylic with a straight handle, this tool is used when reactives are performed directly into cuvettes to mix added reagents. Up and down motion in cell provides complete mixing• Shaft diameter- 3.17mm (0.125 inches)