Learn More
Fireflysci P68 Mixer for Semi-Micro Cuvettes (Type 9, Type 9B, Type 9BM, Type 9F, Type 9M)
SDP
Catalog No. 501060509
Encompass_Preferred
Manual mixer fits any standard 10mm pathlength cuveette with a 4mm inside width• Made of clear acrylic with a straight handle, this tool is used when reactives are performed directly into cuvettes to mix added reagents. Up and down motion in cell provides complete mixing• Shaft diameter- 3.17mm (0.125 inches)
Catalog No. 50-106-0509 Supplier Fireflysci Supplier No. P68
Manual mixer fits any standard 10mm pathlength cuveette with a 4mm inside width• Made of clear acrylic with a straight handle, this tool is used when reactives are performed directly into cuvettes to mix added reagents. Up and down motion in cell provides complete mixing• Shaft diameter- 3.17mm (0.125 inches)