Learn More
Fireflysci Plastic Black Case for 45-55mm Tall Cells- Holds 4 Cells
SDP
Catalog No. 501060490
Encompass_Preferred
Black plastic case for holding either four 10mm lightpath cells, two 20mm lightpath cells or one 50 or 100mm lightpath cell• Can hold up to 45mm in height cuvettes
Catalog No. 50-106-0490 Supplier Fireflysci Supplier No. CASEP4
Black plastic case for holding either four 10mm lightpath cells, two 20mm lightpath cells or one 50 or 100mm lightpath cell• Can hold up to 45mm in height cuvettes