Learn More
Fireflysci P72 Mini Cuvette Stirrer for Semi-Micro Cuvettes (Type 9, Type 9B, Type 9BM, Type 9FL, Type 9M)
SDP
Catalog No. 501060511
Encompass_Preferred
Mini cuvette stirrer for use in semi-micro cuvettes and narrow-apertured cuvettes• This PTFE magnetic centrifugal stirrer fits into cuvettes with smaller apertures and volumes and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirring machine• Centrifugal pumping action generated by the cross channels in the upper face mixers without aeration. Photometric measurements can proceed uninterrupted while reagents are added• Dimensions- 7x2mm• For use with narrow-necked and semi-micro cuvettes• For use in Type 9, Type 9B, Type 9BM, Type 9FL and Type 9M cuvettes
Catalog No. 50-106-0511 Supplier Fireflysci Supplier No. P72
Mini cuvette stirrer for use in semi-micro cuvettes and narrow-apertured cuvettes• This PTFE magnetic centrifugal stirrer fits into cuvettes with smaller apertures and volumes and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirring machine• Centrifugal pumping action generated by the cross channels in the upper face mixers without aeration. Photometric measurements can proceed uninterrupted while reagents are added• Dimensions- 7x2mm• For use with narrow-necked and semi-micro cuvettes• For use in Type 9, Type 9B, Type 9BM, Type 9FL and Type 9M cuvettes