Learn More
Description
- For use on the ATAC 6000 analyzer
- Designed for compatibility with this product
- Acrylonitrile butadiene styrene (ABS) construction
- Natural color
Specifications
Specifications
| Color | Natural |
| Material | ABS |
| No. per Case | 300 |
| Packaging Type | Individually Wrapped |
| Quantity | 300/Cs. |
| For Use With (Equipment) | ATAC 6000 Analyzer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.