Learn More
Description
- For use on the Cobas Mira, Mira S and Mira Plus analyzers
- 12-place cuvette segments
- Individually wrapped
- Polymethylmethacrylate (PMMA)
- Natural color
- Case of 300
Specifications
Specifications
| For Use With (Equipment) | Cobas Mira, Mira S, and Mira Plus Analyzers |
| Color | Natural |
| Quantity | 300/Cs. |
| Material | PMMA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.