Learn More
Mettler Toledo™ Cuvette Holder
Catalog No. p-7229345 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Pathlength:
10/16 mm
20/30 mm
40/50 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pathlength
01-911-523 10/16 mm
01-911-524 20/30 mm
01-911-525 40/50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-911-523 Supplier Mettler Toledo™ Supplier No. 30705122
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mettler Toledo cuvette holders provide a secure base for your sample, ensuring highly accurate measurement results.
  • Compatible with Easy UV and Easy Vis Spectrophotometers
TRUSTED_SUSTAINABILITY
Specifications
Type Cell Holder System
Pathlength 10/16 mm