Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Cuvettes for Horiba ABX Pentra 400

Catalog No. 21200113 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
450/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-113 450/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-113 Supplier Globe Scientific Supplier No. 5162
This item is not returnable. View return policy

Designed specifically for use with the Horiba Pentra 400 chemistry analyzers. Globe Scientific Cuvettes for Horiba ABX Pentra 400 provide a great alternative to OEM cuvettes. Cuvette segments are constructed from polymethylmethacrylate.

  • For use with the Horiba Pentra 400 chemistry analyzers
  • Great alternative to OEM cuvettes
  • Cuvette segments constructed from polymethylmethacrylate (PMMA)
  • Natural color

Specifications

Color Natural
Quantity 450/Cs.
Material PMMA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.