Learn More
Starna Cells Inc CUVETTE PATH LENGTH 1MM
SDP
Catalog No. NC9981513
Encompass_Preferred
Dual Path Length or Fluorimeter, Cell type: Open top, Type: 52, Path Length: 1 or 10, Internal Width: 1, External: 12.5L x 12.5W x 45mm H, Window Material: SOG (Special optical glass 320-2500nm), Q (Far UV quartz 170-2700nm).
Catalog No. NC9981513 Supplier Starna Cells Inc Supplier No. 52Q1
Dual Path Length or Fluorimeter, Cell type: Open top, Type: 52, Path Length: 1 or 10, Internal Width: 1, External: 12.5L x 12.5W x 45mm H, Window Material: SOG (Special optical glass 320-2500nm), Q (Far UV quartz 170-2700nm).