Learn More
MTC Bio Spectrophotometer Cuvette, PS, 1.5mL semi-micro, square opening, 10mm light path, 340-800nm range, 2 optical sides, 10 trays of 100, 1000/cs
Catalog No. NC3737120
Encompass_Preferred
MTC Bio's disposable cuvettes are compatible with most popular spectrophotometers. Manufactured from high quality, optically clear polystyrene. They are intended for deteriminations in the 340 to 800nm (visible) wavelength range. Available in 3 sizes: 1.5mL micro, 2.5mL semi-micro and 4.5mL macro. All sizes are 12.5mm square, 45mm height and are compatible with various spectrophotometer models including: Carolina®, Spectronic®200, Spectronic®20 and 20D, Spectronic®Educator, Genesys™ and Accuris SmartDrop. If a closure is desired, please order cuvette caps separately. • Fits most spectrophotometers and photometers• Polystyrene for use in visible range (340-800nm)• Dust-proof, non-scratch protective packaging• Absorption reproductivity error of ≤1%
Catalog No. NC3737120 Supplier MTC Bio Supplier No. S41003
MTC Bio's disposable cuvettes are compatible with most popular spectrophotometers. Manufactured from high quality, optically clear polystyrene. They are intended for deteriminations in the 340 to 800nm (visible) wavelength range. Available in 3 sizes: 1.5mL micro, 2.5mL semi-micro and 4.5mL macro. All sizes are 12.5mm square, 45mm height and are compatible with various spectrophotometer models including: Carolina®, Spectronic®200, Spectronic®20 and 20D, Spectronic®Educator, Genesys™ and Accuris SmartDrop. If a closure is desired, please order cuvette caps separately. • Fits most spectrophotometers and photometers• Polystyrene for use in visible range (340-800nm)• Dust-proof, non-scratch protective packaging• Absorption reproductivity error of ≤1%