Learn More
World Precision Instrument Micro-volume Flow Cuvettes, 10mm Path
SDP
Catalog No. 50823029
Encompass
  • WPI’s glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments.
  • Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications.
Catalog No. 50-823-029 Supplier World Precision Instrument Supplier No. CUV20611
  • WPI’s glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments.
  • Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications.