Learn More
World Precision Instrument Semi-micro, self-masking Quartz Cuvette, 10mm path, Style D, V=1.0mL
SDP
Catalog No. 50823024
Encompass
WPI’s glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments. Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications, as it does not exhibit background fluorescence. Quartz cuvettes (absorbance, fluorescence and flow) are shipped individually packaged. These economic quartz are ideal for precision measurements because of their high quality materials used and their low manufacturing tolerances.
Catalog No. 50-823-024 Supplier World Precision Instrument Supplier No. CUV20321
 Restricted Item
Add to Cart
Add to Cart
WPI’s glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments. Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications, as it does not exhibit background fluorescence. Quartz cuvettes (absorbance, fluorescence and flow) are shipped individually packaged. These economic quartz are ideal for precision measurements because of their high quality materials used and their low manufacturing tolerances.