Learn More
Andwin Scientific CUVETTE QUARTZ 3.5ML EA
SDP
Catalog No. 501305918
Encompass_Preferred
The quart cuvette has 2 clear sides, 2 frosted sides, a 3.5 mL capacity, and a 10 mm pathlength, making it an ideal tool for routine laboratory testing. With over 80% transmission at 200 nm, this product works well for the entire UV spectra. This product comes with a chemical resistant, PTFE covers.
Catalog No. 50-130-5918 Supplier Andwin Scientific Supplier No. 8330113A
The quart cuvette has 2 clear sides, 2 frosted sides, a 3.5 mL capacity, and a 10 mm pathlength, making it an ideal tool for routine laboratory testing. With over 80% transmission at 200 nm, this product works well for the entire UV spectra. This product comes with a chemical resistant, PTFE covers.