Learn More
Cole-Parmer Cole-Parmer Standard-Sized cuvettes; quartz; 3.5 mL capacity
Catalog No. NC0054436 Shop All Cole-Parmer Products
Encompass_Preferred
Standard-size cuvettes are the type most often used for routine laboratory testing. Cuvettes include chemical resistant, PTFE covers. Cuvettes have two clear sides and two frosted sides. Transmission is over 80% at 200 nm.
Catalog No. NC0054436 Supplier Cole-Parmer Supplier No. 8330113
Standard-size cuvettes are the type most often used for routine laboratory testing. Cuvettes include chemical resistant, PTFE covers. Cuvettes have two clear sides and two frosted sides. Transmission is over 80% at 200 nm.