Learn More
METTLER TOLEDO 3P INDUSTRIAL Cuvette, quartz glass, 10 mm
Catalog No. NC1661083
Encompass
Cuvette, 10 mm pathlength, SUPRASIL quartz glass, 190 - 2500 nm, PTFE lid. Excellent measurement characteristics in the ultra-violet and visible range.
Catalog No. NC1661083 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 30258738
Cuvette, 10 mm pathlength, SUPRASIL quartz glass, 190 - 2500 nm, PTFE lid. Excellent measurement characteristics in the ultra-violet and visible range.