Learn More
Perkin Elmer US LLC Cuvette Rack
Catalog No. 502112924
Encompass_Preferred
The Cuvette Rack holds up to sixteen standard 10 mm cells. It provides easy, durable storage space and has numbered positions to aid sample tracking. It can withstand temperatures up to 121°C and can be used in an autoclave.
Catalog No. 50-211-2924 Supplier Perkin Elmer US LLC Supplier No. L7110275
The Cuvette Rack holds up to sixteen standard 10 mm cells. It provides easy, durable storage space and has numbered positions to aid sample tracking. It can withstand temperatures up to 121°C and can be used in an autoclave.