Learn More
Sarstedt Inc Cuvette, Semi-Micro, 1.6mL, 100/Pack
Catalog No. NC9754913 Shop All Sarstedt Inc. Products
Encompass_Preferred
Semi-micro cuvette, 10 x 4mm, optical pathway 10mm, Length: 45mm, Material: Polystyrene, 100/BX, 2000Pcs./CS
Catalog No. NC9754913 Supplier Sarstedt Inc Supplier No. 67.742
Semi-micro cuvette, 10 x 4mm, optical pathway 10mm, Length: 45mm, Material: Polystyrene, 100/BX, 2000Pcs./CS