Learn More
Chemglass Life Sciences UV-Cuvette, Macro
SDP
Catalog No. 50143748
Encompass_Preferred
UV-Cuvette, Macro. UV-Transparent Cuvettes are a popular disposable alternative to quartz cuvettes in DNA, RNA and protein analysis as well as many other applications between 220nm and 900nm. Choose macro cuvettes for large volume samples or for applications where chemical resistance is primary.UV-Cuvettes eliminate the tedious & time-wasting cuvette washing & the risk of sample carry-over associated with reusable quartz cuvettes. Process QC applications avoid the need for qualified SOP’s for cuvette cleaning.The proprietary plastic material offers chemical compatibility superior to that of conventional plastic cuvettes of acrylic or polystyrene, and may be used with most polar organic solvents.Cuvettes have recessed windows to help protect against scratching and fit most commercial spectrophotometers and photometers. They are grouped by mold cavity number to maximize consistency between cuvettes and reduce variation in extinction coefficient.
Catalog No. 50-143-748 Supplier Chemglass Life Sciences Supplier No. CLS1377001
UV-Cuvette, Macro. UV-Transparent Cuvettes are a popular disposable alternative to quartz cuvettes in DNA, RNA and protein analysis as well as many other applications between 220nm and 900nm. Choose macro cuvettes for large volume samples or for applications where chemical resistance is primary.UV-Cuvettes eliminate the tedious & time-wasting cuvette washing & the risk of sample carry-over associated with reusable quartz cuvettes. Process QC applications avoid the need for qualified SOP’s for cuvette cleaning.The proprietary plastic material offers chemical compatibility superior to that of conventional plastic cuvettes of acrylic or polystyrene, and may be used with most polar organic solvents.Cuvettes have recessed windows to help protect against scratching and fit most commercial spectrophotometers and photometers. They are grouped by mold cavity number to maximize consistency between cuvettes and reduce variation in extinction coefficient.