Learn More
Chemglass Life Sciences UV-Cuvette, Semi-Micro
SDP
Catalog No. 50143749
Encompass_Preferred
UV-Cuvette, Semi-Micro. UV-Transparent Cuvettes are a disposable alternative to quartz cuvettes in DNA, RNA and protein analysis as well as many other applications between 220nm and 900nm. Use the semi-micro size UV-Cuvettes for sample volumes between 1.5 and 3.0mL and the macro UV-Cuvettes for samples larger than 2.5 up to 4.5mL.UV-Cuvettes eliminate the tedious & time-wasting cuvette washing & the risk of sample carry-over associated with reusable quartz cuvettes. Process QC applications avoid the need for qualified SOP’s for cuvette cleaning.The proprietary plastic material offers chemical compatibility superior to that of conventional plastic cuvettes of acrylic or polystyrene, and may be used with most polar organic solvents.Cuvettes have recessed windows to help protect against scratching and fit most commercial spectrophotometers and photometers. They are grouped by mold cavity number to maximize consistency between cuvettes and reduce variation in extinction coefficient.
Catalog No. 50-143-749 Supplier Chemglass Life Sciences Supplier No. CLS1377003
UV-Cuvette, Semi-Micro. UV-Transparent Cuvettes are a disposable alternative to quartz cuvettes in DNA, RNA and protein analysis as well as many other applications between 220nm and 900nm. Use the semi-micro size UV-Cuvettes for sample volumes between 1.5 and 3.0mL and the macro UV-Cuvettes for samples larger than 2.5 up to 4.5mL.UV-Cuvettes eliminate the tedious & time-wasting cuvette washing & the risk of sample carry-over associated with reusable quartz cuvettes. Process QC applications avoid the need for qualified SOP’s for cuvette cleaning.The proprietary plastic material offers chemical compatibility superior to that of conventional plastic cuvettes of acrylic or polystyrene, and may be used with most polar organic solvents.Cuvettes have recessed windows to help protect against scratching and fit most commercial spectrophotometers and photometers. They are grouped by mold cavity number to maximize consistency between cuvettes and reduce variation in extinction coefficient.