Learn More
Sigma Aldrich Fine Chemicals Biosciences Cuvette washer Single1EA
Catalog No. 502900749
Encompass
Supplied with a rubber stopper that fits most filter flasks and with neoprene gaskets that form seals with the open ends of the cuvets.
Catalog No. 50-290-0749 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C12951EA
Supplied with a rubber stopper that fits most filter flasks and with neoprene gaskets that form seals with the open ends of the cuvets.