Learn More
Boca Scientific Inc Electroporation Cuvettes, 1mm Universal Fit with Long Electrode, pack of 50
Catalog No. NC1158126
Encompass
These Electroporation Cuvettes are designed to maximize molecular electroporation and electrofusion efficiencies for Bacteria, Yeast, Insect, Plant and Mammalian cells. These cuvettes are compatible with most electroporation systems. The cap design improves aseptic handling techniques and the positive seal reduces potential aerosol and contamination issues. The molding process ensures extremely fine tolerances so that the electrodes have a consistent gap and parallel configuration. Each batch of cuvettes undergoes rigorous testing at several stages during the manufacturing process for engineering tolerances, biocompatibility and sterility. The cuvettes are supplied individually wrapped.
Catalog No. NC1158126 Supplier Boca Scientific Inc Supplier No. EP201
These Electroporation Cuvettes are designed to maximize molecular electroporation and electrofusion efficiencies for Bacteria, Yeast, Insect, Plant and Mammalian cells. These cuvettes are compatible with most electroporation systems. The cap design improves aseptic handling techniques and the positive seal reduces potential aerosol and contamination issues. The molding process ensures extremely fine tolerances so that the electrodes have a consistent gap and parallel configuration. Each batch of cuvettes undergoes rigorous testing at several stages during the manufacturing process for engineering tolerances, biocompatibility and sterility. The cuvettes are supplied individually wrapped.