Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Cuvettes for Ciba Corning 550 Express

Catalog No. 21200118 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
100/Cs.
200/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-118 200/Cs.
21-200-117 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-200-118 Supplier Globe Scientific Supplier No. 5109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with the Bayer 550 Express & Express Plus analyzers. Globe Scientific Cuvettes for Ciba Corning 550 Express are packaged in cartridges for easy loading into the analyzers. Each cartridge contains 13 (5-place) cuvette segments.

  • For use with the Bayer 550 Express & Express Plus analyzers
  • Cuvettes are packaged in cartridges for easy loading into the analyzers
  • Each cartridge contains 13 (5-place) cuvette segments
  • Polymethylmethacrylate (PMMA)
  • Natural color
  • Available with 24 disposable bags (21-200-117)

Specifications

Type Cuvette
For Use With (Equipment) Bayer 550 Express & Express Plus Analyzers
Color Natural
Includes Cuvettes only
Quantity 200/Cs.
Material PMMA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.