Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Cuvettes for Mindray™ BS300

Catalog No. 21200137 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
1000/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-137 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-137 Supplier Globe Scientific Supplier No. 5146
This item is not returnable. View return policy

Designed for use with Mindray™ BS300 chemistry analyzers. Globe Scientific Cuvettes for Mindray™ BS300 are produced from polystyrene (PS) and are offered individually wrapped in cases of 1000.

  • For use with Mindray™ BS300 chemistry analyzers
  • Cuvette segments are produced from polystyrene (PS)
  • Individually wrapped
  • Natural color

Specifications

Color Natural
Quantity 1000/Cs.
Material Polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.