Learn More
Andwin Scientific CUVETTES PS4.5ML10X10X45 500CS
SDP
Catalog No. NC0092598
Encompass_Preferred
Cuvette Macro For spectrophotometry Polystyrene (PS) 10mm path length mold-cavity matched for minimum variability in extinction cfficient. Minimum fill volume 2.5mL maximum fill volume 4.5mL PK500., 500/CS
Catalog No. NC0092598 Supplier Andwin Scientific Supplier No. 97000584
Cuvette Macro For spectrophotometry Polystyrene (PS) 10mm path length mold-cavity matched for minimum variability in extinction cfficient. Minimum fill volume 2.5mL maximum fill volume 4.5mL PK500., 500/CS