Learn More
Cole-Parmer Semimicro cuvettes with frosted thick wall; cover top; quartz; 1.4 mL capacity
Catalog No. NC9069499 Shop All Cole-Parmer Products
Encompass_Preferred
The glass cuvettes have % transmission guaranteed 82% at 320 nm. Tolerance will be within 1%. The quartz cuvette has 80% transmission guaranteed at 200 nm. Tolerance is within 1%
Catalog No. NC9069499 Supplier Cole-Parmer Supplier No. 8320032
The glass cuvettes have % transmission guaranteed 82% at 320 nm. Tolerance will be within 1%. The quartz cuvette has 80% transmission guaranteed at 200 nm. Tolerance is within 1%