Learn More
Beckman Coulter CUVETTES RECT.QUARTZ 10X2MM 2
Catalog No. NC9760767 Shop All Beckman Coulter Products
Encompass
Starna Rectangular with Quartz to Pyrex graded seal (GS), Path Length: 10 mm, Internal Width: 2 mm, Nominal Volume: 0.700 mL, Micro
Catalog No. NC9760767 Supplier Beckman Coulter Supplier No. 533041
Starna Rectangular with Quartz to Pyrex graded seal (GS), Path Length: 10 mm, Internal Width: 2 mm, Nominal Volume: 0.700 mL, Micro