Learn More
Aviva Systems Biology Corp CUX1 Antibody - N-terminal region (ARP34139_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34139_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CUX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CUX1. Alternate names: CASP, CDP, CDP/Cut, CDP1, COY1, CUTL1, CUX, Clox, Cux/CDP, GOLIM6, Nbla10317, p100, p110, p200, p75ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55266