Learn More
Enzo Life Sciences CV-Caspase-3 & 7 detection kit (1 Kit)
SDP
Catalog No. 502010249
Encompass_Preferred
A consistant feature of mammalian cell apoptosis is the activation of caspases 3 and/or 7. Based on the dye cresyl violet and the caspase 3/7 DEVD peptide recognition sequence, this kit allows the detection of caspases 3 and 7 in living cells by fluorescence microscopy. When cleaved by caspase 3 or 7, the mono and unsubstituted cresyl violet fluoresce when excited at 550-590 nm. Once cells are grown, and apoptosis induced, staining is completed in just a few minutes. Hoechst 33342 and Acridine Orange are also included to allow visualization of nuclei and lysosomes. Applications: Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-0249 Supplier Enzo Life Sciences Supplier No. BMLAK1180001
A consistant feature of mammalian cell apoptosis is the activation of caspases 3 and/or 7. Based on the dye cresyl violet and the caspase 3/7 DEVD peptide recognition sequence, this kit allows the detection of caspases 3 and 7 in living cells by fluorescence microscopy. When cleaved by caspase 3 or 7, the mono and unsubstituted cresyl violet fluoresce when excited at 550-590 nm. Once cells are grown, and apoptosis induced, staining is completed in just a few minutes. Hoechst 33342 and Acridine Orange are also included to allow visualization of nuclei and lysosomes. Applications: Fluorescence microscopy, Fluorescent detection .