Learn More
Enzo Life Sciences CV-Cathepsin L detection kit (1 Kit)
SDP
Catalog No. 502010252
Encompass_Preferred
Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis. Based on the dye cresyl violet and the cathepsin L PheArg peptide recognition sequence, this kit allows the detection of cathepsin L activity in living cells by fluorescence microscopy. Once cells are grown, staining is completed in just a few minutes. Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes. Applications: Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-0252 Supplier Enzo Life Sciences Supplier No. BMLAK1270001
Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis. Based on the dye cresyl violet and the cathepsin L PheArg peptide recognition sequence, this kit allows the detection of cathepsin L activity in living cells by fluorescence microscopy. Once cells are grown, staining is completed in just a few minutes. Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes. Applications: Fluorescence microscopy, Fluorescent detection .