Learn More
Sino Biological Coxsackievirus (CV) A6 VP1 Protein (Fc Tag)
Catalog No. 502993393
Encompass_Preferred
A DNA sequence encoding the Coxsackievirus(CV)A6 VP1 (Q9YLP0) (Asn1-Phe305) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-299-3393 Supplier Sino Biological Supplier No. 41054V02B100
A DNA sequence encoding the Coxsackievirus(CV)A6 VP1 (Q9YLP0) (Asn1-Phe305) was expressed with the Fc region of human IgG1 at the C-terminus.