Learn More
Sino Biological Coxsackievirus (CV) A6 VP1 Protein (His Tag)
Catalog No. 502992975
Encompass_Preferred
A DNA sequence encoding the Coxsackievirus(CV)A6 VP1 (Q9YLP0) (Asn1-Phe305) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2975 Supplier Sino Biological Supplier No. 41054V08B100
A DNA sequence encoding the Coxsackievirus(CV)A6 VP1 (Q9YLP0) (Asn1-Phe305) was expressed with a polyhistidine tag at the C-terminus.