Learn More
New England Biolabs, Inc. CviKI-1 – 250 units
SDP
Catalog No. 50994874
Encompass
A restriction endonuclease that recognizes the sequence RG^_CY. CviKI-1 is a restriction endonuclease with 4 expected recognition sites as well as up to eleven relaxed non-cognate sites (star sites).
Catalog No. 50-994-874 Supplier New England Biolabs, Inc. Supplier No. R0710S
A restriction endonuclease that recognizes the sequence RG^_CY. CviKI-1 is a restriction endonuclease with 4 expected recognition sites as well as up to eleven relaxed non-cognate sites (star sites).