Learn More
New England Biolabs, Inc. CviQI – 10000 units
SDP
Catalog No. 50995261
Encompass
A restriction endonuclease that recognizes the sequence G^TA_C.
Catalog No. 50-995-261 Supplier New England Biolabs, Inc. Supplier No. R0639L
A restriction endonuclease that recognizes the sequence G^TA_C.