Learn More
Medchemexpress LLC HY-134661A 5mg , CVN424 CAS:2254706-21-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134661A5MG
Encompass_Preferred
HY-134661A 5mg CVN424 CAS: 2254706-21-1 CVN424 is an orally active and selective GPR6 inverse agonist with a Ki of 9.4 nM and an EC50 of 38 nM. CVN424 is brain-penetrant and has the potential for Parkinson disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0065