Learn More
BIOSCIENCES TOOLS CVR SLIP GLS 22X22 MM 100/PK
Catalog No. NC0854651
Encompass
Cover glasses for high performance microscopes, made of chemically resistant borosilicate glass of the first hydrolytic class with precision thickness No. 1.5 (0.170 mm +/- 0.005 mm), suitable for in-vitro diagnostic applications.
Catalog No. NC0854651 Supplier BIOSCIENCES TOOLS Supplier No. CSHPNO1.522X22
Cover glasses for high performance microscopes, made of chemically resistant borosilicate glass of the first hydrolytic class with precision thickness No. 1.5 (0.170 mm +/- 0.005 mm), suitable for in-vitro diagnostic applications.