Learn More
Andwin Scientific CVRALL BP SMS LG BLU CS25
SDP
Catalog No. NC0621619
Encompass
Constructed from SMS fabric, the basic protection LG SMS coverall provides light fluid and particulate barrier while remaining comfortable and breathable in high-class environments. It is low-linting and resistant to tears and punctures.
This item is not returnable. View return policy
Catalog No. NC0621619 Supplier Andwin Scientific Supplier No. 414004325
Constructed from SMS fabric, the basic protection LG SMS coverall provides light fluid and particulate barrier while remaining comfortable and breathable in high-class environments. It is low-linting and resistant to tears and punctures.