Learn More
Electron Microscopy Sciences Gold Seal 3419 Cover Glass #1.5, 24 x 30 mm, 94 pieces/oz Packaging 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 6379010
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3419
Size: 24 x 30 mm
Packaging: 94 pieces/oz
Catalog No. 50-189-9136