Learn More
Electron Microscopy Sciences Gold Seal 3422 Cover Glass #1.5, 24 x 50 mm, 58 pieces/oz Packaging 1 OZ
SDP
Catalog No. 501899137
Encompass_Preferred
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3422
Size: 24 x 50 mm
Packaging: 58 pieces/oz
Catalog No. 50-189-9137 Supplier Electron Microscopy Sciences Supplier No. 6379210
Made from pre-selected, pre-cleaned silicate glass. Packaged with desiccants in a lint free box.
Gold Seal: 3422
Size: 24 x 50 mm
Packaging: 58 pieces/oz