Learn More
Electron Microscopy Sciences Platinum Line Coverglass #1.5, 22 x 50 mm 1 OZ
SDP
Catalog No. 501928974
Encompass_Preferred
Platinum Line No. 1.5 Coverglass are made of a water-white colorless borosilicate glass, with an optimum hydrolytic resistance. Ultra-thin. Performs exceptionally well on all automated glass cverslippers.
Thickness: #1.5
Size: 22 x 50 mm
Catalog No. 50-192-8974 Supplier Electron Microscopy Sciences Supplier No. 7188730
Platinum Line No. 1.5 Coverglass are made of a water-white colorless borosilicate glass, with an optimum hydrolytic resistance. Ultra-thin. Performs exceptionally well on all automated glass cverslippers.
Thickness: #1.5
Size: 22 x 50 mm