Learn More
Electron Microscopy Sciences Platinum Line Coverglass #1.5, 24 x 30 mm 1 OZ
SDP
Catalog No. 501928975
Encompass_Preferred
Platinum Line No. 1.5 Coverglass are made of a water-white colorless borosilicate glass, with an optimum hydrolytic resistance. Ultra-thin. Performs exceptionally well on all automated glass cverslippers.
Thickness: #1.5
Size: 24 x 30 mm
Catalog No. 50-192-8975 Supplier Electron Microscopy Sciences Supplier No. 7188731
Platinum Line No. 1.5 Coverglass are made of a water-white colorless borosilicate glass, with an optimum hydrolytic resistance. Ultra-thin. Performs exceptionally well on all automated glass cverslippers.
Thickness: #1.5
Size: 24 x 30 mm