Learn More
Electron Microscopy Sciences Platinum Line Coverglass Round #1, 12 mm 0.5 OZ
SDP
Catalog No. 501928951
Encompass_Preferred
Platinum Line Round Coverglass are made of a water-white colorless borosilicate glass, with an optimuim hydrolytic resistance. These ultra-thin coverglasses perform exceptionally well on all automated glass coverslippers.
Thickness: #1
Diameter: 12 mm
Catalog No. 50-192-8951 Supplier Electron Microscopy Sciences Supplier No. 7188703
Platinum Line Round Coverglass are made of a water-white colorless borosilicate glass, with an optimuim hydrolytic resistance. These ultra-thin coverglasses perform exceptionally well on all automated glass coverslippers.
Thickness: #1
Diameter: 12 mm