Learn More
Electron Microscopy Sciences Platinum Line Coverglass Round #1.5, 18 mm 0.5 OZ
SDP
Supplier:  Electron Microscopy Sciences 7188707
Encompass_Preferred
Platinum Line Round Coverglass are made of a water-white colorless borosilicate glass, with an optimuim hydrolytic resistance. These ultra-thin coverglasses perform exceptionally well on all automated glass coverslippers.
Thickness: #1.5
Diameter: 18 mm
Catalog No. 50-192-8954