Learn More
Electron Microscopy Sciences Platinum Line Coverglass Square #1, 22 x 22 mm 1 OZ
SDP
Catalog No. 501928961
Encompass_Preferred
Platinum Line No. 1 Coverglass are made of a water-white colorless borosilicate glass, with an optimum hydrolytic resistance. Ultra-thin. Performs exceptionally well on all automated glass cverslippers.
Thickness: #1
Size: 22 x 22 mm
Catalog No. 50-192-8961 Supplier Electron Microscopy Sciences Supplier No. 7188714
Platinum Line No. 1 Coverglass are made of a water-white colorless borosilicate glass, with an optimum hydrolytic resistance. Ultra-thin. Performs exceptionally well on all automated glass cverslippers.
Thickness: #1
Size: 22 x 22 mm